accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBrO

CAS RN 21921-76-6

IUPAC Name: 4-bromofuran-2-carbaldehyde
Synonyms: 4-bromofuran-2-carbaldehyde
Molecular Weight (g/mol): 174.98
Molecular Formula: C5H3BrO2
InChi Key: MRGBBKQOSUHKPF-UHFFFAOYSA-N
SMILES: BrC1=COC(C=O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)