accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 219297-13-9

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-4-(pyridin-4-yl)butanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-4-(pyridin-4-yl)butanoic acid
Molecular Weight (g/mol): 280.32
Molecular Formula: C14H20N2O4
InChi Key: BYTKQINZDKLYGV-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(O)=O)CC1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)