accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHNOOOPOOOPOOOOHOHNaNaNa

CAS RN 21931-53-3

IUPAC Name: trisodium ({[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: trisodium {[5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol): 470.11
Molecular Formula: C9H11N2Na3O12P2
InChi Key: GFASEGWJDXDCKZ-UHFFFAOYNA-K
SMILES: [Na+].[Na+].[Na+].OC1C(O)C(OC1COP([O-])(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results