accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 219543-09-6

IUPAC Name: 4-acetamido-2,2,6,6-tetramethyl-1-oxo-1λ⁵-piperidin-1-ylium; tetrafluoroboranuide
Synonyms: 4-acetamido-2,2,6,6-tetramethyl-1-oxo-1λ⁵-piperidin-1-ylium tetrafluoroborate
Molecular Weight (g/mol): 300.1
Molecular Formula: C11H21BF4N2O2
InChi Key: HTMHEICBCHCWAU-UHFFFAOYSA-O
SMILES: F[B-](F)(F)F.CC(=O)NC1CC(C)(C)[N+](=O)C(C)(C)C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)