accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21968-17-2

IUPAC Name: (propan-2-yl)(propyl)amine
Synonyms: isopropyl-propyl-amine
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: VLSTXUUYLIALPB-UHFFFAOYSA-N
SMILES: CCCNC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results