accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2198-53-0

IUPAC Name: N-(2,6-dimethylphenyl)acetamide
Synonyms: 2,6-dimethylacetanilide
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: NRPTXWYBRKRZES-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C(C)C=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results