accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2199-44-2

IUPAC Name: ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
Synonyms: ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: IZSBSZYFPYIJDI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C=C(C)N1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)