accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2199-51-1

IUPAC Name: ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate
Synonyms: ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: QWSFDUPEOPMXCV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)NC=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results