accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2199-58-8

IUPAC Name: 3,5-dimethyl-1H-pyrrole-2-carbaldehyde
Synonyms: 3,5-dimethyl-1H-pyrrole-2-carbaldehyde
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: RDFZYUOHJBXMJA-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N1)C=O
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (4)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (1)
  • (7)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)