accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHCH3OH3C

CAS RN 2199-64-6

IUPAC Name: ethyl 4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
Synonyms: ethyl 4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: CLJUICOFPKFFGJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C(C=O)=C(C)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results