accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2199-86-2

IUPAC Name: 6,8-dichloro-2-oxo-2H-chromene-3-carboxylic acid
Synonyms: 6,8-dichloro-2-oxochromene-3-carboxylic acid
Molecular Weight (g/mol): 259.04
Molecular Formula: C10H4Cl2O4
InChi Key: WJEHZKIVHXRVSR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC(Cl)=CC(Cl)=C2OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results