accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 219967-69-8

IUPAC Name: (2S)-4-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (2S)-4-[4-(tert-butoxy)phenyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 473.57
Molecular Formula: C29H31NO5
InChi Key: FKUBPXDMQFEPGP-CYLJNIGPNA-N
SMILES: CC(C)(C)OC1=CC=C(CC[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results