CAS RN 220088-42-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Binucleine 2
A cell-permeable pyrazolo compound that inhibits cytokinesis function of the Aurora B kinase pathway.