CAS RN 22009-38-7
IUPAC Name:
7-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms:
7-hydroxy-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol):
162.19
Molecular Formula:
C10H10O2
InChi Key:
LGFSAJZSDNYVCW-UHFFFAOYSA-N
SMILES:
OC1=CC2=C(CCCC2=O)C=C1
1
–
1
of
1
results
1
7-Hydroxy-1-tetralone 98.0+%, TCI America™
CAS: 22009-38-7 Molecular Formula: C10H10O2 Molecular Weight (g/mol): 162.188 MDL Number: MFCD01312225 InChI Key: LGFSAJZSDNYVCW-UHFFFAOYSA-N Synonym: 7-Hydroxy-3,4-dihydronaphthalen-1(2H)-one PubChem CID: 571818 IUPAC Name: 7-hydroxy-3,4-dihydro-2H-naphthalen-1-one SMILES: C1CC2=C(C=C(C=C2)O)C(=O)C1