CAS RN 220191-36-6
CAS RN 220191-36-6
IUPAC Name:
3,4-bis(2,4,5-trimethylthiophen-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms:
3,4-bis(2,4,5-trimethylthiophen-3-yl)-1H-pyrrole-2,5-dione
Molecular Weight (g/mol):
345.48
Molecular Formula:
C18H19NO2S2
InChi Key:
OHZCQTZIDIVCPI-UHFFFAOYSA-N
SMILES:
CC1=C(C)C(=C(C)S1)C1=C(C(=O)NC1=O)C1=C(C)SC(C)=C1C
1
–
1
of
1
results
1
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide 97.0+%, TCI America™
CAS: 220191-36-6 Molecular Formula: C18H19NO2S2 Molecular Weight (g/mol): 345.48 MDL Number: MFCD00142790 InChI Key: OHZCQTZIDIVCPI-UHFFFAOYSA-N PubChem CID: 22023748 IUPAC Name: 3,4-bis(2,4,5-trimethylthiophen-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione SMILES: CC1=C(C)C(=C(C)S1)C1=C(C(=O)NC1=O)C1=C(C)SC(C)=C1C