accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrOO

CAS RN 220212-12-4

IUPAC Name: benzyl 3-bromopyrrolidine-1-carboxylate
Synonyms: benzyl 3-bromopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 284.15
Molecular Formula: C12H14BrNO2
InChi Key: LWEKWWHCQBWMQC-UHFFFAOYNA-N
SMILES: BrC1CCN(C1)C(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)