accessibility menu, dialog, popup

UserName

Viewing profile as user:
FSOOClFF

CAS RN 220227-21-4

IUPAC Name: 2,4,5-trifluorobenzene-1-sulfonyl chloride
Synonyms: 2,4,5-trifluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 230.59
Molecular Formula: C6H2ClF3O2S
InChi Key: STCZWXXRRTWRSK-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C=C1F)S(Cl)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results