accessibility menu, dialog, popup

UserName

Viewing profile as user:
FSOOClFF

CAS RN 220239-64-5

IUPAC Name: 2,4,6-trifluorobenzene-1-sulfonyl chloride
Synonyms: 2,4,6-trifluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 230.59
Molecular Formula: C6H2ClF3O2S
InChi Key: XINCBNCLCIQIJM-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C(F)=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)