CAS RN 22027-52-7
IUPAC Name:
methyl 4-(3-ethoxy-3-oxopropanoyl)benzoate
Synonyms:
methyl 4-(3-ethoxy-3-oxopropanoyl)benzoate
Molecular Weight (g/mol):
250.25
Molecular Formula:
C13H14O5
InChi Key:
GFDCYWYWNSTMDX-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC
1
–
1
of
1
results
1
Promotions Available
Methyl 4-methoxycarbonylbenzoylacetate, 95%, Thermo Scientific™
Methyl 4-methoxycarbonylbenzoylacetate, 95%, Thermo Scientific™
CAS: 22027-52-7 Molecular Formula: C13H14O5 Molecular Weight (g/mol): 250.25 MDL Number: MFCD00800637 InChI Key: GFDCYWYWNSTMDX-UHFFFAOYSA-N PubChem CID: 3864207 IUPAC Name: methyl 4-(3-methoxy-3-oxopropanoyl)benzoate SMILES: CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC