accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 220290-68-6

IUPAC Name: 2-oxa-6-azaspiro[3.4]octane
Synonyms: 2-oxa-6-azaspiro[3.4]octane
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: ZHAIMJRKJKQNQI-UHFFFAOYSA-N
SMILES: C1OCC11CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results