accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 220298-96-4

IUPAC Name: tert-butyl N-[4-(aminomethyl)phenyl]carbamate
Synonyms: tert-butyl N-[4-(aminomethyl)phenyl]carbamate
Molecular Weight (g/mol): 222.29
Molecular Formula: C12H18N2O2
InChi Key: URXUHALBOWYXJZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results