accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22031-12-5

IUPAC Name: [3-(diphenylphosphanyl)-2-[(diphenylphosphanyl)methyl]-2-methylpropyl]diphenylphosphane
Synonyms: [3-(diphenylphosphanyl)-2-[(diphenylphosphanyl)methyl]-2-methylpropyl]diphenylphosphane
Molecular Weight (g/mol): 624.68
Molecular Formula: C41H39P3
InChi Key: BARUNXKDFNLHEV-UHFFFAOYSA-N
SMILES: CC(CP(C1=CC=CC=C1)C1=CC=CC=C1)(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results