CAS RN 22034-43-1
IUPAC Name:
9-(naphthalen-1-yl)-9H-carbazole
Synonyms:
9-(naphthalen-1-yl)carbazole
Molecular Weight (g/mol):
293.37
Molecular Formula:
C22H15N
InChi Key:
QSOAYCUFEQGHDN-UHFFFAOYSA-N
SMILES:
C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC2=CC=CC=C12
1
–
1
of
1
results
1
9-(1-Naphthyl)carbazole 98.0+%, TCI America™
CAS: 22034-43-1 Molecular Formula: C22H15N Molecular Weight (g/mol): 293.37 MDL Number: MFCD26127424 InChI Key: QSOAYCUFEQGHDN-UHFFFAOYSA-N PubChem CID: 626218 IUPAC Name: 9-(naphthalen-1-yl)-9H-carbazole SMILES: C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC2=CC=CC=C12