accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOO

CAS RN 22042-73-5

IUPAC Name: 4-(2-hydroxyethoxy)benzaldehyde
Synonyms: 4-(2-hydroxyethoxy)benzaldehyde
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: VCDGTEZSUNFOKA-UHFFFAOYSA-N
SMILES: OCCOC1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results