accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22042-79-1

IUPAC Name: 3,5-dimethyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Synonyms: 3,5-dimethyl-1-phenylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: VOMRTQQGXWPTJK-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1C=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results