accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 220497-79-0

HOO(R)BrNHOO

CAS RN 220497-79-0

IUPAC Name: (2R)-3-(2-bromophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2R)-3-(2-bromophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 466.33
Molecular Formula: C24H20BrNO4
InChi Key: GRJPAUULVKPBHU-JOCHJYFZSA-N
SMILES: OC(=O)[C@@H](CC1=CC=CC=C1Br)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)