accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONHOOHO

CAS RN 220497-85-8

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(furan-2-yl)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(furan-2-yl)propanoic acid|FMOC-D-2-FURYLALANINE
Molecular Weight (g/mol): 377.4
Molecular Formula: C22H19NO5
InChi Key: AJXDCHXGNUFBRC-UHFFFAOYSA-N
SMILES: OC(=O)C(CC1=CC=CO1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)