accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(S)(S)OH(S)HO(R)HO

CAS RN 220497-88-1

IUPAC Name: (1S,2R,3S,4S)-2,3-dihydroxy-4-(hydroxymethyl)cyclopentan-1-aminium
Synonyms: (1S,2R,3S,4S)-2,3-dihydroxy-4-(hydroxymethyl)cyclopentan-1-aminium
Molecular Weight (g/mol): 148.18
Molecular Formula: C6H14NO3
InChi Key: FHNKBDPGQXLKRW-OMMKOOBNSA-O
SMILES: [NH3+][C@H]1C[C@@H](CO)[C@H](O)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results