accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22053-74-3

H3CSO

CAS RN 22053-74-3

IUPAC Name: 3-methyl-1-benzothiophene-2-carbaldehyde
Synonyms: 3-methyl-1-benzothiophene-2-carbaldehyde
Molecular Weight (g/mol): 176.23
Molecular Formula: C10H8OS
InChi Key: DRZGHNXLEQHVHB-UHFFFAOYSA-N
SMILES: CC1=C(SC2=CC=CC=C12)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results