accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)(S)H3C

CAS RN 2207-03-6

IUPAC Name: (1S,3S)-1,3-dimethylcyclohexane
Synonyms: (1S,3S)-1,3-dimethylcyclohexane
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: SGVUHPSBDNVHKL-YUMQZZPRSA-N
SMILES: C[C@H]1CCC[C@H](C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results