accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3

CAS RN 2207-04-7

IUPAC Name: 1,4-dimethylcyclohexane
Synonyms: 1,4-dimethylcyclohexane 1,4-dimethylcyclohexane (cis)
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: QRMPKOFEUHIBNM-UHFFFAOYSA-N
SMILES: CC1CCC(C)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)