accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HONHNNHNNH2ClNNCH3H3C

CAS RN 220792-57-4

IUPAC Name: 2-({6-[(3-amino-5-chlorophenyl)amino]-9-(propan-2-yl)-9H-purin-2-yl}amino)-3-methylbutan-1-ol
Synonyms: 2-({6-[(3-amino-5-chlorophenyl)amino]-9-isopropylpurin-2-yl}amino)-3-methylbutan-1-ol
Molecular Weight (g/mol): 403.92
Molecular Formula: C19H26ClN7O
InChi Key: RAMROQQYRRQPDL-UHFFFAOYNA-N
SMILES: CC(C)C(CO)NC1=NC(NC2=CC(Cl)=CC(N)=C2)=C2N=CN(C(C)C)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results