accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2208-05-1

IUPAC Name: 2-(dimethylamino)ethyl benzoate
Synonyms: benzoic acid,dimethylaminoethyl ester
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: KJSGODDTWRXQRH-UHFFFAOYSA-N
SMILES: CN(C)CCOC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results