accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22082-93-5

IUPAC Name: 1-bromo-2-phenylnaphthalene
Synonyms: 1-bromo-2-phenylnaphthalene
Molecular Weight (g/mol): 283.17
Molecular Formula: C16H11Br
InChi Key: ANKXTSUKPPHQJR-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=CC=C1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results