accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOO

CAS RN 22084-89-5

IUPAC Name: 3-(2-methylphenyl)propanoic acid
Synonyms: 3-(2-methylphenyl)propanoic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: JIRKNEAMPYVPTD-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1CCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results