CAS RN 22099-23-6
IUPAC Name:
8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-4-carbaldehyde
Synonyms:
8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-4-carbaldehyde
Molecular Weight (g/mol):
212.27
Molecular Formula:
C13H8OS
InChi Key:
QFGXAEFJJINHAI-UHFFFAOYSA-N
SMILES:
O=CC1=CC2=C(SC3=C2C=CC=C3)C=C1
1
–
1
of
1
results
1
Dibenzothiophene-2-carboxaldehyde 98.0+%, TCI America™
CAS: 22099-23-6 Molecular Formula: C13H8OS Molecular Weight (g/mol): 212.266 MDL Number: MFCD02251670 InChI Key: QFGXAEFJJINHAI-UHFFFAOYSA-N PubChem CID: 641360 IUPAC Name: dibenzothiophene-2-carbaldehyde SMILES: C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)C=O