accessibility menu, dialog, popup

UserName

Viewing profile as user:
BIHOOH

CAS RN 221037-98-5

IUPAC Name: (3-iodophenyl)boronic acid
Synonyms: 3-iodophenylboronic acid
Molecular Weight (g/mol): 247.83
Molecular Formula: C6H6BIO2
InChi Key: REEUXWXIMNEIIN-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(I)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results