CAS RN 221044-05-9
IUPAC Name:
1-(pyrimidin-2-yl)-1H-indole
Synonyms:
1-(pyrimidin-2-yl)indole
Molecular Weight (g/mol):
195.23
Molecular Formula:
C12H9N3
InChi Key:
CNAQMKUJWYJTRY-UHFFFAOYSA-N
SMILES:
C1=CC2=CC=CC=C2N1C1=NC=CC=N1
1
–
1
of
1
results
1
1-(2-Pyrimidinyl)-1H-indole 98.0+%, TCI America™
CAS: 221044-05-9 Molecular Formula: C12H9N3 Molecular Weight (g/mol): 195.225 MDL Number: MFCD28556906 InChI Key: CNAQMKUJWYJTRY-UHFFFAOYSA-N PubChem CID: 53381231 IUPAC Name: 1-pyrimidin-2-ylindole SMILES: C1=CC=C2C(=C1)C=CN2C3=NC=CC=N3