accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNNH

CAS RN 22112-89-6

IUPAC Name: 2,12-diphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Synonyms: 2,12-diphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Molecular Weight (g/mol): 462.56
Molecular Formula: C32H22N4
InChi Key: QIBKIAFNCVIIMG-UHFFFAOYSA-N
SMILES: N1C2=CC=C1C(=C1C=CC(C=C3C=CC(=N3)C(=C3NC(C=C3)=C2)C2=CC=CC=C2)=N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results