accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSON

CAS RN 2212-67-1

IUPAC Name: (azepan-1-yl)(ethylsulfanyl)methanone
Synonyms: jalan stauffer R-4,572 caswell no. 444 molinate (iso) C9H17nos malerbane giavoni L felan hydram higalnate ialan
Molecular Weight (g/mol): 187.3
Molecular Formula: C9H17NOS
InChi Key: DEDOPGXGGQYYMW-UHFFFAOYSA-N
SMILES: CCSC(=O)N1CCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results