accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2212-69-3

IUPAC Name: 4-nitrophenyl (2S)-2-{[(benzyloxy)carbonyl]amino}-6-{[(tert-butoxy)carbonyl]amino}hexanoate
Synonyms: 4-nitrophenyl (2S)-2-{[(benzyloxy)carbonyl]amino}-6-[(tert-butoxycarbonyl)amino]hexanoate|Z-LYS(BOC)-ONP
Molecular Weight (g/mol): 501.54
Molecular Formula: C25H31N3O8
InChi Key: XBMRVOHLFIZDDV-NRFANRHFSA-N
SMILES: CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)