accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22122-36-7

IUPAC Name: 3-methyl-2,5-dihydrofuran-2-one
Synonyms: 3-methyl-5H-furan-2-one
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: VGHBEMPMIVEGJP-UHFFFAOYSA-N
SMILES: CC1=CCOC1=O
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)