accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22123-14-4

IUPAC Name: 2-chloro-6-methyl-4-(trifluoromethyl)pyridine
Synonyms: 2-chloro-6-methyl-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 195.57
Molecular Formula: C7H5ClF3N
InChi Key: SXLBWNSGCIEART-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=CC(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results