accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22123-19-9

IUPAC Name: 6-methyl-4-(trifluoromethyl)-1,2-dihydropyridin-2-one
Synonyms: 6-methyl-4-(trifluoromethyl)-1H-pyridin-2-one
Molecular Weight (g/mol): 177.13
Molecular Formula: C7H6F3NO
InChi Key: CDRMKCNOGIBXOG-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(=O)N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results