accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2213-43-6

IUPAC Name: piperidin-1-amine
Synonyms: N-aminopiperidine
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: LWMPFIOTEAXAGV-UHFFFAOYSA-N
SMILES: NN1CCCCC1
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)