accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 221347-71-3

IUPAC Name: 5-fluoro-1,3-benzoxazole
Synonyms: 5-fluoro-1,3-benzoxazole
Molecular Weight (g/mol): 137.11
Molecular Formula: C7H4FNO
InChi Key: ZRMPAEOUOPNNPZ-UHFFFAOYSA-N
SMILES: FC1=CC=C2OC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results