accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 221352-82-5

IUPAC Name: 2-[4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)piperidin-1-yl]acetic acid
Synonyms: (4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}piperidin-1-yl)acetic acid
Molecular Weight (g/mol): 380.44
Molecular Formula: C22H24N2O4
InChi Key: VLQJIWJYRZIBMP-UHFFFAOYSA-N
SMILES: OC(=O)CN1CCC(CC1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)