accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBr

CAS RN 22148-20-5

IUPAC Name: 1-(4-bromophenyl)piperidine
Synonyms: 1-(4-bromophenyl)piperidine
Molecular Weight (g/mol): 240.14
Molecular Formula: C11H14BrN
InChi Key: OACWQRGNHICZFD-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1CCCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)