accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2215-76-1

IUPAC Name: 4-(4-formylphenoxy)benzaldehyde
Synonyms: 4-(4-formylphenoxy)benzaldehyde
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: GXZZHLULZRMUQC-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)