accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2216-52-6

IUPAC Name: 5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Synonyms: menthol natural caswell no. 540 menthol racemic menthol preparation brazilian menthol natural (1α,2β,5α)-menthol cis-1,3,trans-1,4-menthol menthol, (1α,2β,5α)-isomer (1α,2β,5α)-isomer menthol d-neomenthol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: NOOLISFMXDJSKH-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results